Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pigu | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004721.1 | NP_001004721.1 | 12 | 1584 | ||
GenBank mRNA | AK143517.1 | - | 12 | 1584 | ||
AK171988.1 | - | 12 | 1563 | |||
BC013076.1 | - | 6 | 5407 | |||
BC028278.1 | - | 4 | 715 |