Search Thermo Fisher Scientific
- Order Status
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: OR6Q1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005186.2 | NP_001005186.2 | 1 | 486 | ||
GenBank mRNA | BC151146.1 | - | 1 | 509 | 9021017 | |
BC151147.1 | - | 1 | 509 | 9021018 |