Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Txlna | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005506.3 | NP_001005506.2 | 5 | 1199 | ||
NM_001199695.1 | NP_001186624.1 | 6 | 916 | |||
GenBank mRNA | AK135290.1 | - | 5 | 1047 | ||
AK141252.1 | - | 6 | 850 | |||
AK150240.1 | - | 6 | 873 | |||
AK160829.1 | - | 5 | 1047 | |||
AK161649.1 | - | 6 | 879 | |||
AK168418.1 | - | 5 | 1200 | |||
AK171101.1 | - | 6 | 892 | |||
BC060227.1 | - | 5 | 1034 | 30357541 | ||
BC065115.1 | - | 5 | 1034 | 30362350 |