Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tm9sf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005554.1 | NP_001005554.1 | 15 | 1820 | ||
GenBank mRNA | BC081873.1 | - | 15 | 1820 | 7107003 | |
FQ209454.1 | - | 3 | 248 |