Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ptbp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005555.1 | NP_001005555.1 | 3 | 135 | ||
XM_006233236.2 | XP_006233298.1 | 3 | 669 | |||
XM_006233237.2 | XP_006233299.1 | 3 | 668 | |||
XM_006233238.2 | XP_006233300.1 | 3 | 668 | |||
GenBank mRNA | AJ010585.1 | - | 3 | 104 | ||
BC082076.1 | - | 3 | 135 | 7133221 |