Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PITRM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242307.1 | NP_001229236.1 | 14 | 1591 | ||
NM_001242309.1 | NP_001229238.1 | 13 | 1488 | |||
NM_014889.3 | NP_055704.2 | 14 | 1591 | |||
XM_005252345.3 | XP_005252402.1 | 14 | 1591 | |||
XM_017015470.1 | XP_016870959.1 | 14 | 1659 | |||
XM_017015471.1 | XP_016870960.1 | 14 | 1659 | |||
XM_017015472.1 | XP_016870961.1 | 14 | 1488 | |||
XM_017015473.1 | XP_016870962.1 | 14 | 1488 | |||
GenBank mRNA | AB029027.2 | - | 4 | 320 | ||
AF061243.1 | - | 14 | 1535 | |||
AK002061.1 | - | 14 | 1559 | |||
AK225084.1 | - | 14 | 1529 | |||
AK294865.1 | - | 13 | 1504 | |||
AK299464.1 | - | 13 | 1337 | |||
AK303406.1 | - | 13 | 1429 | |||
AK315117.1 | - | 14 | 1529 | |||
BC001150.1 | - | 8 | 891 | 3503609 | ||
BC005025.1 | - | 14 | 1526 | 4040225 | ||
BC045577.1 | - | 14 | 2001 | |||
BC095422.1 | - | 14 | 1532 | 4822631 | ||
BC111987.1 | - | 14 | 1547 | 8327455 | ||
BC113369.1 | - | 14 | 1547 | 8322421 | ||
BX649017.1 | - | 14 | 1533 | |||
CR749279.1 | - | 12 | 1267 |