Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: AGAP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037131.2 | NP_001032208.1 | 3, 4 | 916 | ||
NM_001244888.1 | NP_001231817.1 | 3, 4 | 916 | |||
NM_014914.4 | NP_055729.2 | 3, 4 | 916 | |||
XM_006712235.3 | XP_006712298.1 | 3, 4 | 506 | |||
XM_006712237.3 | XP_006712300.2 | 3, 4 | 1163 | |||
GenBank mRNA | AB029022.1 | - | 3, 4 | 889 | ||
AF413078.1 | - | 3, 4 | 318 | |||
AY033765.1 | - | 3, 4 | 879 | |||
BC060814.1 | - | 3, 4 | 916 | |||
BC140856.1 | - | 4, 5 | 533 | 9021724 | ||
BC167153.1 | - | 3, 4 | 889 | 8862550 | ||
BC167157.1 | - | 3, 4 | 901 | 8862571 |