Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other PHACTR1 SIRNA Assays ›
Gene Symbol:
PHACTR1
Gene Name:
phosphatase and actin regulator 1
Gene Aliases:
RPEL, RPEL1, dJ257A7.2
Chromosome Location:
Chr.6: 12716635 - 13290484 on Build GRCh38
UniGene ID:
Hs.436996
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
221447
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: PHACTR1
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242648.2
NP_001229577.1
9
1552
NM_001322308.1
NP_001309237.1
8
1507
NM_001322309.1
NP_001309238.1
8
1398
NM_001322310.1
NP_001309239.1
8
1473
NM_001322311.1
NP_001309240.1
6
1159
NM_001322312.1
NP_001309241.1
6
1522
NM_001322313.1
NP_001309242.1
7
1729
NM_001322314.1
NP_001309243.1
7
1661
NM_030948.3
NP_112210.1
9
1742
XM_005248934.2
XP_005248991.1
10
1791
XM_017010452.1
XP_016865941.1
7
1455
XM_017010454.1
XP_016865943.1
10
1791
XM_017010455.1
XP_016865944.1
10
1759
XM_017010456.1
XP_016865945.1
9
1756
XM_017010457.1
XP_016865946.1
9
1735
XM_017010458.1
XP_016865947.1
6
1248
XM_017010459.1
XP_016865948.1
9
1584
XM_017010460.1
XP_016865949.1
9
1552
XM_017010461.1
XP_016865950.1
8
1523
XM_017010462.1
XP_016865951.1
8
1549
XM_017010463.1
XP_016865952.1
7
1649
XM_017010464.1
XP_016865953.1
6
1248
XM_017010465.1
XP_016865954.1
7
1429
XM_017010466.1
XP_016865955.1
7
1442
XM_017010467.1
XP_016865956.1
6
1429
XM_017010469.1
XP_016865958.1
6
1231
GenBank mRNA
AB051520.1
-
8
1349
AK290017.1
-
9
1403
AK295263.1
-
10
1759
BC101526.1
-
8
1316
8069032
BC101528.1
-
8
1316
8069034
Back To Top
More Information
Panther Classification:
Molecular Function -
phosphatase modulator
Gene Ontology Categories:
Function(s)
Process(es)
actomyosin structure organization
actin cytoskeleton reorganization
negative regulation of catalytic activity
stress fiber assembly
cell motility
actin binding
protein phosphatase inhibitor activity
protein phosphatase 1 binding
protein phosphatase type 1 regulator activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-r4vqj:80/10.239.163.135:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0