Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Icmt | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_133788.2 | NP_598549.1 | 5 | 4318 | ||
GenBank mRNA | AF209926.1 | - | 5 | 4100 | ||
AK154410.1 | - | 6 | 4342 | |||
BC006724.1 | - | 1 | 767 | |||
BC057132.1 | - | 6 | 4337 | |||
BC110695.1 | - | 5 | 4305 | 6842587 |