Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Krt5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183333.1 | NP_899162.1 | 427 | |||
GenBank mRNA | AY342389.1 | - | 427 | |||
BC062086.1 | - | 290 | 5599969 |
Gene Symbol: LOC102551453 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008765863.1 | XP_008764085.1 | 1 | 315 |