Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SEC24D | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001318066.1 | NP_001304995.1 | 4 | 633 | ||
NM_014822.3 | NP_055637.2 | 4 | 633 | |||
XM_005263379.2 | XP_005263436.1 | 4 | 507 | |||
GenBank mRNA | AB018298.1 | - | 4 | 536 | ||
AF130464.2 | - | 3 | 336 | |||
AK291765.1 | - | 4 | 533 | |||
AK302479.1 | - | 4 | 427 | |||
BC035761.1 | - | 4 | 555 | 5554774 | ||
BC037736.1 | - | 5 | 677 |