Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TMCC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001242925.1 | NP_001229854.1 | 3 | 1085 | ||
NM_001297611.1 | NP_001284540.1 | 2 | 543 | |||
NM_001297613.1 | NP_001284542.1 | 2 | 563 | |||
NM_014858.3 | NP_055673.2 | 3 | 1749 | |||
XM_005245684.1 | XP_005245741.1 | 3 | 1440 | |||
XM_005245685.4 | XP_005245742.1 | 3 | 1071 | |||
XM_005245686.3 | XP_005245743.1 | 3 | 1466 | |||
XM_017003000.1 | XP_016858489.1 | 2 | 524 | |||
GenBank mRNA | AB001596.1 | - | 2 | 419 | ||
AB007950.3 | - | 3 | 1749 | |||
AK095816.1 | - | 2 | 563 | |||
AK131419.1 | - | 4 | 1538 | |||
AK293734.1 | - | 3 | 1085 | |||
AK314398.1 | - | 2 | 442 | |||
AY358192.1 | - | 2 | 1657 | |||
BC041019.1 | - | 2 | 483 | |||
BC131629.1 | - | 3 | 1221 | 40118729 | ||
BQ222504.1 | - | 3 | 630 |