Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr318 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000505.1 | NP_001000505.1 | 1 | 916 |
Gene Symbol: Olr1874 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_002728868.4 | XP_002728914.2 | 1 | 1521 |