Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olr1657 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001000536.1 | NP_001000536.1 | 1 | 25 |
Gene Symbol: LOC102557552 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771774.1 | XP_008769996.1 | 1 | 78 |
Gene Symbol: LOC103690297 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008771775.1 | XP_008769997.1 | 1 | 42 |