Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CHSY1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014918.4 | NP_055733.2 | 2 | 853 | ||
XM_011521364.2 | XP_011519666.1 | 2 | 392 | |||
GenBank mRNA | AB023207.1 | - | 2 | 871 | ||
AB071402.1 | - | 2 | 391 | |||
AY358529.1 | - | 2 | 865 | |||
BC046247.1 | - | 2 | 860 | 5751618 |