Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZC3H11A SIRNA Assays ›
Gene Symbol:
ZC3H11A
LOC441155
Gene Name:
zinc finger CCCH-type containing 11A
zinc finger CCCH-type domain-containing-like
Gene Aliases:
ZC3HDC11A
-
Chromosome Location:
Chr.1: 203795537 - 203854128 on Build GRCh38
Chromosome Location:
Chr.6: 65301418 - 65306069 on Build GRCh38
UniGene ID:
Hs.532399
Hs.546686
Species:
Human
Species Specific ID (Flybase ID):
-
-
siRNA ID
22259
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZC3H11A
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001319238.1
NP_001306167.1
20
4484
NM_001319239.1
NP_001306168.1
20
4379
NM_014827.5
NP_055642.3
20
4055
XM_005245643.2
XP_005245700.1
20
4074
XM_005245644.3
XP_005245701.1
19
3758
XM_006711673.2
XP_006711736.1
20
3921
XM_011510199.1
XP_011508501.1
18
5126
XM_011510200.1
XP_011508502.1
19
4146
XM_011510201.1
XP_011508503.1
20
4332
XM_011510202.1
XP_011508504.1
18
4071
XM_017002956.1
XP_016858445.1
19
5380
XM_017002957.1
XP_016858446.1
19
5232
XM_017002958.1
XP_016858447.1
19
5219
XM_017002959.1
XP_016858448.1
20
4292
XM_017002960.1
XP_016858449.1
19
5374
XM_017002961.1
XP_016858450.1
18
5120
XM_017002962.1
XP_016858451.1
19
4140
XM_017002963.1
XP_016858452.1
20
3932
XM_017002964.1
XP_016858453.1
19
5292
XM_017002965.1
XP_016858454.1
18
4615
GenBank mRNA
AB014563.1
-
17
3566
AL049419.1
-
1
752
BC038513.2
-
20
4210
BX649148.1
-
17
7691
CR627439.1
-
19
5076
CR627446.1
-
17
7179
Gene Symbol: LOC441155
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
XM_017010889.1
XP_016866378.1
2
3605
Back To Top
More Information
Panther Classification:
Molecular Function -
RNA processing factor
Gene Ontology Categories:
Function(s)
Process(es)
termination of RNA polymerase II transcription
RNA export from nucleus
mRNA export from nucleus
poly(A)+ mRNA export from nucleus
mRNA 3'-end processing
protein binding
poly(A) RNA binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7c5f844ff9-f2nf4:80/10.239.165.207:80.
git-commit: 1d07a45fbd3445ee17db916b8ffd5a90ab36fdd0
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.11.0-2024.07.28-1.0