Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other SNAP91 SIRNA Assays ›
Gene Symbol:
SNAP91
Gene Name:
synaptosome associated protein 91
Gene Aliases:
AP180, CALM
Chromosome Location:
Chr.6: 83552885 - 83709408 on Build GRCh38
UniGene ID:
Hs.368046
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22266
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: SNAP91
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242792.1
NP_001229721.1
30
3771
NM_001242793.1
NP_001229722.1
28
3480
NM_001242794.1
NP_001229723.1
20
2649
NM_001256717.1
NP_001243646.1
30
3838
NM_001256718.1
NP_001243647.1
29
3466
NM_014841.2
NP_055656.1
30
3774
XM_005248770.4
XP_005248827.1
30
3613
XM_006715615.1
XP_006715678.1
30
3565
XM_011536265.1
XP_011534567.1
30
3572
XM_011536266.1
XP_011534568.1
30
3684
XM_011536269.1
XP_011534571.1
30
3550
XM_011536271.1
XP_011534573.1
29
3523
XM_011536273.2
XP_011534575.1
29
3487
XM_011536275.2
XP_011534577.1
29
3481
XM_011536276.2
XP_011534578.1
29
3457
XM_011536277.1
XP_011534579.1
32
3853
XM_017011554.1
XP_016867043.1
29
3723
XM_017011555.1
XP_016867044.1
29
3607
XM_017011556.1
XP_016867045.1
29
3539
XM_017011557.1
XP_016867046.1
30
3727
XM_017011558.1
XP_016867047.1
30
3557
XM_017011559.1
XP_016867048.1
30
3656
XM_017011560.1
XP_016867049.1
30
3598
XM_017011561.1
XP_016867050.1
30
3669
XM_017011562.1
XP_016867051.1
29
3727
XM_017011563.1
XP_016867052.1
28
3721
XM_017011564.1
XP_016867053.1
28
3592
XM_017011565.1
XP_016867054.1
28
3650
XM_017011566.1
XP_016867055.1
29
3700
XM_017011567.1
XP_016867056.1
29
3503
XM_017011568.1
XP_016867057.1
28
3694
XM_017011569.1
XP_016867058.1
29
3685
XM_017011570.1
XP_016867059.1
29
3664
XM_017011571.1
XP_016867060.1
29
3535
XM_017011572.1
XP_016867061.1
29
3494
XM_017011573.1
XP_016867062.1
29
3593
XM_017011574.1
XP_016867063.1
29
3639
XM_017011575.1
XP_016867064.1
29
3658
XM_017011576.1
XP_016867065.1
29
3461
XM_017011577.1
XP_016867066.1
29
3587
XM_017011578.1
XP_016867067.1
29
3600
XM_017011579.1
XP_016867068.1
29
3643
XM_017011580.1
XP_016867069.1
29
3634
XM_017011581.1
XP_016867070.1
29
3464
XM_017011582.1
XP_016867071.1
29
3493
XM_017011583.1
XP_016867072.1
29
3606
XM_017011584.1
XP_016867073.1
29
3449
XM_017011585.1
XP_016867074.1
29
3490
XM_017011586.1
XP_016867075.1
29
3442
XM_017011587.1
XP_016867076.1
29
3548
XM_017011588.1
XP_016867077.1
29
3561
XM_017011589.1
XP_016867078.1
28
3592
XM_017011590.1
XP_016867079.1
28
3421
GenBank mRNA
AB014556.1
-
30
3700
AF054993.1
-
5
1078
CR749348.1
-
30
3805
Back To Top
More Information
Panther Classification:
Molecular Function -
vesicle coat protein
Gene Ontology Categories:
Function(s)
Process(es)
chemical synaptic transmission
protein transport
clathrin coat assembly
regulation of clathrin-mediated endocytosis
protein binding
1-phosphatidylinositol binding
protein kinase binding
clathrin binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-green-cccc65c97-qtpg5:80/10.239.161.70:80.
git-commit: 2b810224bbfb944e11adb8d8c7a69f36ca15064f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.1-offline