Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SUN1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130965.2 | NP_001124437.1 | 14 | 1635 | ||
NM_001171944.1 | NP_001165415.1 | 12 | 1385 | |||
NM_025154.5 | NP_079430.3 | 13 | 1498 | |||
GenBank mRNA | AB018353.1 | - | 15 | 1733 | ||
AB648918.1 | - | 16 | 1924 | |||
AF202724.1 | - | 5 | 534 | |||
AK022816.1 | - | 13 | 1490 | |||
AK025737.1 | - | 11 | 1160 | |||
AK074320.1 | - | 1 | 82 | |||
AK302896.1 | - | 12 | 1357 | |||
AK304150.1 | - | 10 | 1026 | |||
BC142707.1 | - | 14 | 1645 | 40148817 | ||
BC152418.1 | - | 14 | 1733 | 8862528 | ||
BX640866.1 | - | 5 | 1724 | |||
BX648893.1 | - | 5 | 1724 | |||
BX649069.1 | - | 12 | 1390 |