Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LOC100286922 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_037694.1 | 1 | 268 | |||
NR_037695.1 | 1 | 268 | ||||
GenBank mRNA | BC066346.1 | - | 1 | 253 |
Gene Symbol: DNAJB3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001001394.3 | NP_001001394.1 | 1 | 291 | ||
GenBank mRNA | AY083665.1 | - | 1 | 291 | ||
BC017590.2 | - | 1 | 291 | 4828023 | ||
BC024013.2 | - | 1 | 254 | 4826480 |