Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPFIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AB020704.1 | - | 12 | 1929 | ||
AK124535.1 | - | 5 | 2870 | |||
AK126185.1 | - | 1 | 2036 | |||
AK126624.1 | - | 17 | 3368 | |||
EF428334.1 | - | 35 | 4667 |