Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPFIA4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304331.1 | NP_001291260.1 | 30 | 5468 | ||
NM_001304332.1 | NP_001291261.1 | 29 | 5441 | |||
XM_006711586.2 | XP_006711649.1 | 30 | 5359 | |||
XM_011510070.2 | XP_011508372.1 | 18 | 3388 | |||
XM_017002606.1 | XP_016858095.1 | 29 | 5332 | |||
XM_017002607.1 | XP_016858096.1 | 29 | 5224 | |||
XM_017002609.1 | XP_016858098.1 | 29 | 5363 | |||
GenBank mRNA | AB020704.1 | - | 12 | 2597 | ||
AK124535.1 | - | 5 | 3538 | |||
AK126185.1 | - | 1 | 2704 | |||
AK126624.1 | - | 17 | 4036 | |||
EF428334.1 | - | 35 | 5335 |