Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: KIAA0907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014949.3 | NP_055764.2 | 2 | 297 | ||
GenBank mRNA | AB020714.1 | - | 2 | 254 | ||
AK225714.1 | - | 2 | 268 | |||
AK289902.1 | - | 2 | 240 | |||
AY112680.1 | - | 2 | 228 | |||
AY112681.1 | - | 2 | 228 | |||
AY112682.1 | - | 2 | 228 | |||
BC027182.1 | - | 2 | 253 | |||
BC062637.1 | - | 2 | 252 | 5497581 | ||
BI560023.1 | - | 2 | 307 | |||
KU178555.1 | - | 2 | 228 | |||
KU178556.1 | - | 2 | 228 |