Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: SBNO2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001100122.1 | NP_001093592.1 | 2 | 211 | ||
NM_014963.2 | NP_055778.2 | 5 | 543 | |||
XM_011527804.2 | XP_011526106.1 | 5 | 516 | |||
GenBank mRNA | AB023180.1 | - | 5 | 520 | ||
AK125139.1 | - | 2 | 158 | |||
AK292407.1 | - | 5 | 543 | |||
AK296849.1 | - | 2 | 158 | |||
AK300981.1 | - | 6 | 483 | |||
AK309167.1 | - | 2 | 158 | |||
BP315965.1 | - | 2 | 211 |