Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Brd2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212495.1 | NP_997660.1 | 6 | 1722 | ||
XM_006256097.2 | XP_006256159.1 | 7 | 2395 | |||
XM_008772719.1 | XP_008770941.1 | 6 | 1092 | |||
GenBank mRNA | FQ225945.1 | - | 2 | 307 | ||
FQ226688.1 | - | 4 | 702 | |||
FQ232599.1 | - | 3 | 578 |
Gene Symbol: LOC100909544 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_006256037.2 | XP_006256099.1 | 6 | 2301 | ||
XM_008772772.1 | XP_008770994.1 | 6 | 1091 | |||
XM_008772773.1 | XP_008770995.1 | 7 | 2393 |