Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nhp2l1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212515.2 | NP_997680.1 | 3 | 458 | ||
GenBank mRNA | BC058493.1 | - | 3 | 461 | 6887059 | |
FQ220734.1 | - | 459 | ||||
FQ233358.1 | - | 459 |
Gene Symbol: LOC100359574 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008760591.1 | XP_008758813.1 | 1 | 1151 |