Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcb7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_212518.1 | NP_997683.1 | 11 | 1470 | ||
GenBank mRNA | AJ621255.1 | - | 11 | 1470 |