Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other MYCBP2 SIRNA Assays ›
Gene Symbol:
MYCBP2
Gene Name:
MYC binding protein 2, E3 ubiquitin protein ligase
Gene Aliases:
Myc-bp2, PAM, Phr
Chromosome Location:
Chr.13: 77044657 - 77327066 on Build GRCh38
UniGene ID:
Hs.591221
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22390
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: MYCBP2
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_015057.4
NP_055872.4
1
511
XM_005266299.2
XP_005266356.1
1
535
XM_006719779.2
XP_006719842.1
1
535
XM_006719780.2
XP_006719843.1
1
535
XM_006719781.2
XP_006719844.1
1
535
XM_006719782.2
XP_006719845.1
1
535
XM_006719783.2
XP_006719846.1
1
535
XM_006719784.2
XP_006719847.1
1
535
XM_006719785.2
XP_006719848.1
1
535
XM_006719786.2
XP_006719849.1
1
535
XM_006719787.2
XP_006719850.1
1
535
XM_006719788.2
XP_006719851.1
1
535
XM_006719789.2
XP_006719852.1
1
535
XM_017020454.1
XP_016875943.1
1
535
XM_017020455.1
XP_016875944.1
1
535
XM_017020456.1
XP_016875945.1
1
535
XM_017020457.1
XP_016875946.1
1
535
XM_017020458.1
XP_016875947.1
1
535
XM_017020459.1
XP_016875948.1
1
535
XM_017020460.1
XP_016875949.1
1
535
XM_017020461.1
XP_016875950.1
1
535
XM_017020462.1
XP_016875951.1
1
535
XM_017020463.1
XP_016875952.1
1
535
XM_017020464.1
XP_016875953.1
1
535
XM_017020465.1
XP_016875954.1
1
535
XM_017020466.1
XP_016875955.1
1
535
XM_017020467.1
XP_016875956.1
1
535
GenBank mRNA
AF075587.1
-
2
276
Back To Top
More Information
Panther Classification:
Molecular Function -
ubiquitin-protein ligase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
protein ubiquitination
protein binding
zinc ion binding
ligase activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-b9vj9:80/10.239.164.80:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0