Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: 4931440F15Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176829.2 | NP_789799.1 | 1 | 2518 | ||
GenBank mRNA | AK029918.1 | - | 1 | 2518 | ||
BC139107.1 | - | 1 | 2375 | 8862129 | ||
BC139123.1 | - | 1 | 2375 | 8862145 |