Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: R3hdm1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181750.2 | NP_861415.2 | 21 | 2755 | ||
GenBank mRNA | AK122191.1 | - | 2 | 4394 | ||
BC032160.1 | - | 12 | 1646 | |||
BC137772.1 | - | 19 | 2398 | 8860791 | ||
BC144899.1 | - | 19 | 2398 |