Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cdk10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_194444.2 | NP_919426.2 | 13 | 1360 | ||
NM_194446.2 | NP_919428.1 | 13 | 1398 | |||
GenBank mRNA | AK133918.1 | - | 13 | 1353 | ||
AK144801.1 | - | 13 | 1370 | |||
AK170594.1 | - | 13 | 1394 | |||
BC017131.1 | - | 13 | 1335 | 4506458 | ||
BC023736.1 | - | 13 | 1379 | |||
BC052096.1 | - | 8 | 2253 | |||
DQ288857.1 | - | 13 | 1397 |