Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Commd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198732.2 | NP_942027.1 | 8 | 767 | ||
GenBank mRNA | BC060591.1 | - | 8 | 770 | 6920278 |