Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Tpd52l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_198744.2 | NP_942039.1 | 3 | 431 | ||
XM_006235754.2 | XP_006235816.1 | 3 | 430 | |||
XM_006235757.2 | XP_006235819.1 | 3 | 450 | |||
XM_006235759.2 | XP_006235821.1 | 3 | 423 | |||
XM_006235760.2 | XP_006235822.1 | 3 | 445 | |||
XM_006235761.2 | XP_006235823.1 | 3 | 428 | |||
XM_008762499.1 | XP_008760721.1 | 3 | 364 | |||
XM_008762500.1 | XP_008760722.1 | 3 | 362 | |||
GenBank mRNA | BC059167.1 | - | 3 | 432 | 6890828 |