Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other CEP164 SIRNA Assays ›
Gene Symbol:
CEP164
Gene Name:
centrosomal protein 164
Gene Aliases:
NPHP15
Chromosome Location:
Chr.11: 117316346 - 117413266 on Build GRCh38
UniGene ID:
Hs.504009
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
224131
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: CEP164
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271933.1
NP_001258862.1
18
2546
NM_014956.4
NP_055771.4
19
2603
XM_005271453.1
XP_005271510.1
20
4560
XM_005271456.1
XP_005271513.1
19
2874
XM_005271457.1
XP_005271514.1
19
2883
XM_006718788.1
XP_006718851.1
20
4569
XM_006718794.1
XP_006718857.1
18
2796
XM_011542674.1
XP_011540976.1
20
4620
XM_017017364.1
XP_016872853.1
20
4487
XM_017017365.1
XP_016872854.1
20
4639
XM_017017366.1
XP_016872855.1
20
4422
XM_017017367.1
XP_016872856.1
19
4548
XM_017017368.1
XP_016872857.1
20
4620
XM_017017369.1
XP_016872858.1
20
4620
XM_017017370.1
XP_016872859.1
20
4620
XM_017017371.1
XP_016872860.1
19
4542
XM_017017372.1
XP_016872861.1
19
4482
XM_017017373.1
XP_016872862.1
19
4420
XM_017017374.1
XP_016872863.1
19
4215
XM_017017375.1
XP_016872864.1
18
4344
XM_017017376.1
XP_016872865.1
19
4473
XM_017017377.1
XP_016872866.1
21
4579
XM_017017378.1
XP_016872867.1
18
4263
XM_017017379.1
XP_016872868.1
19
2934
XM_017017380.1
XP_016872869.1
18
2596
XM_017017381.1
XP_016872870.1
18
2856
XM_017017382.1
XP_016872871.1
17
2518
XM_017017383.1
XP_016872872.1
18
2785
XM_017017384.1
XP_016872873.1
18
2584
XM_017017385.1
XP_016872874.1
16
2340
XM_017017386.1
XP_016872875.1
16
2500
GenBank mRNA
AB028975.1
-
18
2817
AK125995.1
-
14
2572
AK303067.1
-
17
2457
AL137265.1
-
12
1780
BX647583.1
-
5
3338
BX648682.1
-
19
2603
Back To Top
More Information
Panther Classification:
Molecular Function -
cytoskeletal protein
microtubule or microtubule-binding cytoskeletal protein
Gene Ontology Categories:
Function(s)
Process(es)
G2/M transition of mitotic cell cycle
DNA repair
mitotic nuclear division
cilium assembly
cell division
protein binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-574f4745d9-tbml7:80/10.239.166.229:80.
git-commit: d20c60ba4e48417b3d41b4fa5195ecf3ea66aacf
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.12.0-2024.07.30-1.0