Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP2R3B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013239.4 | NP_037371.2 | 9 | 1376 | ||
XM_011545630.2 | XP_011543932.1 | 9 | 1376 | |||
GenBank mRNA | AB209137.1 | - | 5 | 982 | ||
AF135016.1 | - | 8 | 740 | |||
AF155098.1 | - | 3 | 151 | |||
AL157501.1 | - | 1 | 87 | |||
BC009032.1 | - | 1 | 333 | |||
BC011180.1 | - | 3 | 153 | |||
BC063429.1 | - | 9 | 1131 | 5239974 | ||
BF530463.1 | - | 1 | 344 | |||
BM715684.1 | - | 3 | 116 | |||
BU845905.1 | - | 1 | 539 |