- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Gene Symbol: PHF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290259.1 | NP_001277188.1 | 2 | 478 | ||
NM_001290260.1 | NP_001277189.1 | 2 | 471 | |||
NM_015153.3 | NP_055968.1 | 1 | 130 | |||
XM_005248701.3 | XP_005248758.1 | 2 | 465 | |||
XM_006715426.3 | XP_006715489.1 | 2 | 690 | |||
XM_011535648.2 | XP_011533950.1 | 2 | 347 | |||
XM_017010626.1 | XP_016866115.1 | 2 | 465 | |||
GenBank mRNA | AF091622.1 | - | 1 | 131 | ||
AK055678.1 | - | 2 | 386 | |||
AK290491.1 | - | 2 | 157 | |||
BC113650.1 | - | 1 | 131 | 8322702 | ||
BC113652.1 | - | 1 | 131 | 8322704 | ||
BX648268.1 | - | 2 | 164 | |||
D87685.2 | - | 1 | 119 |