Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SPEN | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015001.2 | NP_055816.2 | 3 | 631 | ||
GenBank mRNA | AF356524.1 | - | 3 | 631 |