Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NUMB | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001005743.1 | NP_001005743.1 | 9 | 952 | ||
NM_001005744.1 | NP_001005744.1 | 9 | 952 | |||
NM_001005745.1 | NP_001005745.1 | 8 | 919 | |||
NM_001320114.1 | NP_001307043.1 | 9 | 972 | |||
NM_003744.5 | NP_003735.3 | 8 | 919 | |||
GenBank mRNA | AF015040.1 | - | 9 | 910 | ||
AF108092.1 | - | 7 | 737 | |||
AF171938.1 | - | 9 | 902 | |||
AF171939.1 | - | 9 | 902 | |||
AF171940.1 | - | 8 | 869 | |||
AF171941.1 | - | 8 | 869 | |||
AK023670.1 | - | 3 | 989 | |||
AK226069.1 | - | 3 | 2488 | |||
AK304193.1 | - | 7 | 755 | |||
AK315130.1 | - | 7 | 800 | |||
BC020788.1 | - | 7 | 800 | |||
BC068476.1 | - | 8 | 919 | 4822300 | ||
BX248073.1 | - | 8 | 809 | |||
BX248775.1 | - | 7 | 773 | |||
BX248781.1 | - | 7 | 814 | |||
BX647137.1 | - | 9 | 911 | |||
EU265734.1 | - | 7 | 671 | |||
EU265735.1 | - | 6 | 638 | |||
EU265736.1 | - | 7 | 671 | |||
EU265737.1 | - | 6 | 638 |