Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ANKRD6 SIRNA Assays ›
Gene Symbol:
ANKRD6
Gene Name:
ankyrin repeat domain 6
Gene Aliases:
-
Chromosome Location:
Chr.6: 89433178 - 89633834 on Build GRCh38
UniGene ID:
Hs.702213
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
22535
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ANKRD6
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001242809.1
NP_001229738.1
13
1669
NM_001242811.1
NP_001229740.1
13
2052
NM_001242813.1
NP_001229742.1
12
1564
NM_001242814.1
NP_001229743.1
10
1290
NM_014942.4
NP_055757.3
13
1669
XM_005248679.3
XP_005248736.1
12
1537
XM_005248680.3
XP_005248737.1
14
3841
XM_005248681.2
XP_005248738.1
13
1770
XM_005248682.3
XP_005248739.1
11
1459
XM_005248683.3
XP_005248740.1
10
1339
XM_005248684.3
XP_005248741.1
7
1057
XM_011535596.2
XP_011533898.1
11
1438
XM_011535597.2
XP_011533899.1
11
1438
XM_011535598.2
XP_011533900.1
12
1539
XM_011535599.2
XP_011533901.1
12
1539
XM_011535600.2
XP_011533902.1
11
1461
XM_017010493.1
XP_016865982.1
11
1432
XM_017010494.1
XP_016865983.1
11
1438
XM_017010495.1
XP_016865984.1
12
1539
XM_017010496.1
XP_016865985.1
12
1703
XM_017010497.1
XP_016865986.1
11
1372
XM_017010498.1
XP_016865987.1
12
1533
XM_017010499.1
XP_016865988.1
10
1333
XM_017010500.1
XP_016865989.1
10
1339
XM_017010501.1
XP_016865990.1
11
1440
XM_017010502.1
XP_016865991.1
11
3121
XM_017010503.1
XP_016865992.1
11
3125
XM_017010504.1
XP_016865993.1
10
1267
XM_017010505.1
XP_016865994.1
11
1434
XM_017010506.1
XP_016865995.1
10
1273
XM_017010507.1
XP_016865996.1
10
1273
XM_017010508.1
XP_016865997.1
10
3017
XM_017010509.1
XP_016865998.1
9
1168
XM_017010510.1
XP_016865999.1
9
1223
XM_017010511.1
XP_016866000.1
9
2926
XM_017010512.1
XP_016866001.1
8
1158
XM_017010513.1
XP_016866002.1
8
1290
XM_017010514.1
XP_016866003.1
7
1059
XM_017010515.1
XP_016866004.1
13
1554
GenBank mRNA
AB023174.1
-
12
1391
AK074189.1
-
4
592
AK092235.1
-
4
1244
AK096867.1
-
10
1152
AK295731.1
-
13
1969
AK307792.1
-
13
1638
AL117504.1
-
10
1070
BC001078.2
-
1
106
BC042173.1
-
13
1634
5454509
Back To Top
More Information
Panther Classification:
Molecular Function -
protein-binding activity modulator
kinase inhibitor
kinase modulator
non-receptor serine/threonine protein kinase
ubiquitin-protein ligase
protein modifying enzyme
Gene Ontology Categories:
Function(s)
positive regulation of JNK cascade
negative regulation of canonical Wnt signaling pathway
positive regulation of Wnt signaling pathway, planar cell polarity pathway
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-5d97d974fc-l9dfq:80/10.239.161.188:80.
git-commit: fe8b04579249ff98075bad6061a6e2a4739e01ce
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.13.0-2024.08.32-1.0