Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ARHGEF9 SIRNA Assays ›
Gene Symbol:
ARHGEF9
Gene Name:
Cdc42 guanine nucleotide exchange factor 9
Gene Aliases:
COLLYBISTIN, EIEE8, HPEM-2, PEM-2, PEM2
Chromosome Location:
Chr.X: 63634967 - 63786025 on Build GRCh38
UniGene ID:
Hs.54697
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
225632
Purification
HPLC
Standard
In-Vivo Ready
Size
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
5 nmol
20 nmol
20 nmol
Cy3 | 20 nmol
FAM-MGB | 20 nmol
40 nmol
40 nmol
100 nmol
250 nmol
250 nmol
1 umol
10 umol
Availability
Inventoried
Catalog #
AM16708
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ARHGEF9
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001173479.1
NP_001166950.1
9
2682
NM_001173480.1
NP_001166951.1
9
3088
NM_015185.2
NP_056000.1
10
3279
XM_005262249.2
XP_005262306.1
10
3341
XM_005262250.3
XP_005262307.1
10
3239
XM_005262251.1
XP_005262308.1
10
2589
XM_005262252.1
XP_005262309.1
11
2608
XM_011530890.1
XP_011529192.1
11
3023
XM_011530892.1
XP_011529194.1
10
2847
XM_017029363.1
XP_016884852.1
11
3368
XM_017029364.1
XP_016884853.1
12
3145
XM_017029365.1
XP_016884854.1
10
3246
XM_017029366.1
XP_016884855.1
11
3027
XM_017029367.1
XP_016884856.1
11
3016
XM_017029368.1
XP_016884857.1
11
3317
XM_017029369.1
XP_016884858.1
9
3209
XM_017029370.1
XP_016884859.1
9
3118
XM_017029371.1
XP_016884860.1
9
3107
XM_017029372.1
XP_016884861.1
9
2457
XM_017029373.1
XP_016884862.1
11
3382
XM_017029374.1
XP_016884863.1
8
2938
XM_017029375.1
XP_016884864.1
9
3003
XM_017029377.1
XP_016884866.1
9
3006
XM_017029379.1
XP_016884868.1
9
2345
XM_017029380.1
XP_016884869.1
8
2826
GenBank mRNA
AB007884.1
-
10
3241
Back To Top
More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Panther Classification:
Molecular Function -
guanyl-nucleotide exchange factor
Gene Ontology Categories:
Function(s)
Process(es)
ion transmembrane transport
regulation of Rho protein signal transduction
positive regulation of apoptotic process
positive regulation of GTPase activity
regulation of small GTPase mediated signal transduction
guanyl-nucleotide exchange factor activity
Rho guanyl-nucleotide exchange factor activity
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-648b49bbc4-8tvtn:80/10.239.165.222:80.
git-commit: cd89e1917689fc5870ae005717bc737506e58003
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.17.2-2024.10.99-1.0