Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC35D1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015139.2 | NP_055954.1 | 12 | 1558 | ||
XM_011541070.1 | XP_011539372.1 | 13 | 1639 | |||
GenBank mRNA | AB044343.1 | - | 12 | 1206 | ||
AK289800.1 | - | 12 | 1556 | |||
D87449.1 | - | 12 | 1260 |