Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RRP12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145114.1 | NP_001138586.1 | 2 | 290 | ||
NM_001284337.1 | NP_001271266.1 | 2 | 290 | |||
NM_015179.3 | NP_055994.2 | 2 | 290 | |||
XM_011539555.1 | XP_011537857.1 | 2 | 243 | |||
XM_011539556.2 | XP_011537858.1 | 2 | 260 | |||
GenBank mRNA | AB014590.1 | - | 2 | 203 | ||
AK022496.1 | - | 2 | 261 | |||
AK296315.1 | - | 2 | 154 | |||
BC012745.2 | - | 2 | 228 | 4025242 | ||
DA227012.1 | - | 2 | 260 | |||
DC417040.1 | - | 2 | 290 |