Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: NOTCH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_017617.4 | NP_060087.3 | 34 | 8894 | ||
XM_011518717.2 | XP_011517019.2 | 31 | 8186 | |||
GenBank mRNA | AB209873.1 | - | 26 | 7431 | ||
AK000012.1 | - | 1 | 1070 | |||
BC013208.1 | - | 4 | 3079 | |||
BC046127.1 | - | 1 | 645 | |||
CR457221.1 | - | 1 | 397 |