Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: VSTM5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144871.1 | NP_001138343.1 | 2 | 199 | ||
GenBank mRNA | DY655155.1 | - | 1 | 48 |