Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQSEC3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001170738.1 | NP_001164209.1 | 14 | 6549 | ||
XM_011520961.1 | XP_011519263.1 | 13 | 6052 | |||
GenBank mRNA | AB029033.1 | - | 12 | 5117 |