Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134476.1 | NP_001127948.1 | 7 | 1051 | ||
NM_015350.2 | NP_056165.1 | 5 | 1108 | |||
XM_005270701.4 | XP_005270758.1 | 5 | 977 | |||
XM_005270702.4 | XP_005270759.1 | 6 | 998 | |||
XM_005270703.4 | XP_005270760.1 | 5 | 2745 | |||
XM_011541143.2 | XP_011539445.1 | 6 | 1035 | |||
XM_011541144.2 | XP_011539446.1 | 7 | 1347 | |||
XM_011541145.2 | XP_011539447.1 | 8 | 1372 | |||
XM_011541146.2 | XP_011539448.1 | 6 | 2802 | |||
XM_011541148.2 | XP_011539450.1 | 6 | 1023 | |||
XM_017000883.1 | XP_016856372.1 | 6 | 1997 | |||
XM_017000884.1 | XP_016856373.1 | 5 | 2899 | |||
XM_017000885.1 | XP_016856374.1 | 5 | 1069 | |||
XM_017000886.1 | XP_016856375.1 | 5 | 973 | |||
GenBank mRNA | AF385436.1 | - | 5 | 1108 | ||
AY358112.1 | - | 4 | 859 | |||
AY640232.1 | - | 3 | 691 | |||
BC030607.2 | - | 7 | 1287 | 4824652 | ||
BC060782.1 | - | 7 | 993 | 5298560 | ||
D86984.2 | - | 5 | 1414 |