Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CC2D1B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032449.2 | NP_115825.1 | 12 | 1450 | ||
XM_005270590.2 | XP_005270647.1 | 12 | 1536 | |||
XM_005270591.2 | XP_005270648.1 | 12 | 1536 | |||
XM_005270593.2 | XP_005270650.1 | 8 | 1272 | |||
XM_006710423.2 | XP_006710486.1 | 12 | 1536 | |||
XM_017000559.1 | XP_016856048.1 | 12 | 1466 | |||
GenBank mRNA | AB058739.1 | - | 8 | 881 | ||
BC014080.2 | - | 8 | 902 | |||
BC016880.1 | - | 6 | 659 | 3913149 | ||
BC039308.1 | - | 11 | 1860 |