Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SASH1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015278.3 | NP_056093.3 | 9 | 1262 | ||
XM_005266875.1 | XP_005266932.1 | 2 | 488 | |||
XM_011535640.1 | XP_011533942.1 | 9 | 705 | |||
XM_017010598.1 | XP_016866087.1 | 9 | 1494 | |||
XM_017010599.1 | XP_016866088.1 | 9 | 817 | |||
XM_017010600.1 | XP_016866089.1 | 9 | 705 | |||
XM_017010601.1 | XP_016866090.1 | 8 | 919 | |||
XM_017010602.1 | XP_016866091.1 | 7 | 616 | |||
XM_017010603.1 | XP_016866092.1 | 7 | 555 | |||
XM_017010604.1 | XP_016866093.1 | 6 | 464 | |||
XM_017010605.1 | XP_016866094.1 | 6 | 430 | |||
XM_017010606.1 | XP_016866095.1 | 2 | 488 | |||
XM_017010607.1 | XP_016866096.1 | 10 | 885 | |||
GenBank mRNA | AB018333.1 | - | 9 | 1005 | ||
AJ507735.1 | - | 9 | 793 | |||
AK024403.1 | - | 3 | 247 | |||
AK025495.1 | - | 9 | 1262 | |||
AK300403.1 | - | 2 | 172 | |||
AY927559.1 | - | 2 | 108 | |||
BC063279.1 | - | 9 | 731 |