Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SAMM50 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015380.4 | NP_056195.3 | 3, 4 | 439 | ||
GenBank mRNA | AF151809.1 | - | 3, 4 | 351 | ||
AK001087.1 | - | 3, 4 | 378 | |||
AK222657.1 | - | 3, 4 | 313 | |||
AK297770.1 | - | 3, 4 | 274 | |||
AK312773.1 | - | 3, 4 | 274 | |||
AY189688.1 | - | 3, 4 | 354 | |||
BC007830.1 | - | 3, 4 | 346 | 3635175 | ||
BC011681.2 | - | 3, 4 | 399 | 3010765 | ||
BC015200.1 | - | 3, 4 | 289 | 4333051 | ||
CR456483.1 | - | 3, 4 | 251 | |||
DB030637.1 | - | 3, 4 | 439 |