Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: UBR2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015255.2 | NP_056070.1 | 43 | 5062 | ||
XM_005248965.4 | XP_005249022.1 | 43 | 5179 | |||
XM_005248966.3 | XP_005249023.1 | 42 | 4462 | |||
XM_011514438.2 | XP_011512740.2 | 43 | 5262 | |||
XM_011514439.1 | XP_011512741.1 | 34 | 3629 | |||
XM_011514440.1 | XP_011512742.1 | 34 | 3629 | |||
XM_011514441.2 | XP_011512743.1 | 31 | 3242 | |||
XM_017010594.1 | XP_016866083.1 | 43 | 5263 | |||
XM_017010595.1 | XP_016866084.1 | 43 | 5260 | |||
XM_017010596.1 | XP_016866085.1 | 43 | 5094 | |||
GenBank mRNA | AB002347.1 | - | 31 | 3324 | ||
AK026998.1 | - | 22 | 2325 | |||
AK125795.1 | - | 34 | 3603 | |||
AK127310.1 | - | 7 | 1232 | |||
AK304070.1 | - | 35 | 3715 | |||
AY061884.1 | - | 43 | 4764 | |||
BX647467.1 | - | 25 | 3718 | |||
HM370397.1 | - | 1 | 72 |