Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: LRRC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001134476.1 | NP_001127948.1 | 7 | 1973 | ||
NM_015350.2 | NP_056165.1 | 5 | 2030 | |||
XM_005270701.4 | XP_005270758.1 | 5 | 1899 | |||
XM_005270702.4 | XP_005270759.1 | 6 | 1920 | |||
XM_005270703.4 | XP_005270760.1 | 5 | 3667 | |||
XM_011541143.2 | XP_011539445.1 | 6 | 1957 | |||
XM_011541144.2 | XP_011539446.1 | 7 | 2269 | |||
XM_011541145.2 | XP_011539447.1 | 8 | 2294 | |||
XM_011541146.2 | XP_011539448.1 | 6 | 3724 | |||
XM_011541148.2 | XP_011539450.1 | 6 | 1945 | |||
XM_017000883.1 | XP_016856372.1 | 6 | 2919 | |||
XM_017000884.1 | XP_016856373.1 | 5 | 3821 | |||
XM_017000885.1 | XP_016856374.1 | 5 | 1991 | |||
XM_017000886.1 | XP_016856375.1 | 5 | 1895 | |||
GenBank mRNA | AF385436.1 | - | 5 | 2030 | ||
AY358112.1 | - | 4 | 1781 | |||
AY640232.1 | - | 3 | 1611 | |||
BC030607.2 | - | 7 | 2209 | 4824652 | ||
BC060782.1 | - | 7 | 1915 | 5298560 | ||
D86984.2 | - | 5 | 2336 |